Learn More
4-Chloropyridine-2-carboxylic acid, 95%, Thermo Scientific™
Product Code. 15274348 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15274348 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15274348 Supplier Thermo Scientific Acros Supplier No. 454430010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 5470-22-4
Melting Point 180.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C6H4ClNO2
Quantity 1 g
Synonym 4-chloropicolinic acid, 4-chloro-pyridine-2-carboxylic acid, 4-chloro-2-pyridinecarboxylic acid, 4-chloro-2-picolinic acid, 2-pyridinecarboxylic acid, 4-chloro, 4-chloro picolinic acid, 2-carboxy-4-chloropyridine, chloropicolinicacid, pubchem12731, acmc-1bn7z
InChI Key NNMYRMGMVLMQAY-UHFFFAOYSA-N
SMILES C1=CN=C(C=C1Cl)C(=O)O
IUPAC Name 4-chloropyridine-2-carboxylic acid
Molecular Weight (g/mol) 157.56
PubChem CID 230890
Formula Weight 157.56
Percent Purity 95%
Chemical Name or Material 4-Chloropyridine-2-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloropyridine-2-carboxylic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.