Learn More
4-Chloro Loratadine, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 4-(4,8-Dichloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidine-1-carboxylate |
| CAS | 165739-83-3 |
| Molecular Formula | C22 H22 Cl2 N2 O2 |
| Quantity | 1 mg |
| Synonym | Ethyl 4-(4,8-Dichloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidine-1-carboxylate, Loratadine Imp. C (EP) |
| InChI Formula | InChI=1S/C22H22Cl2N2O2/c1-2-28-22(27)26-11-8-14(9-12-26)20-17-6-4-16(23)13-15(17)3-5-18-19(24)7-10-25-21(18)20/h4,6-7,10,13H,2-3,5,8-9,11-12H2,1H3 |
| SMILES | CCOC(=O)N1CCC(=C2c3ccc(Cl)cc3CCc4c(Cl)ccnc24)CC1 |
| IUPAC Name | ethyl 4-(4,8-dichloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate |
| Molecular Weight (g/mol) | 417.33 |
| Formula Weight | 416.1058 |