Learn More
4-Chloro-6-ethyl-5-fluoropyrimidine, TRC
Product Code. 30352726
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30352726 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30352726 Supplier Toronto Research Chemicals Supplier No. TRCC36609525G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Chloro-6-ethyl-5-fluoropyrimidine
CAS 137234-74-3
Molecular Formula C6 H6 Cl F N2
Quantity 25 g
Synonym 4-Chloro-6-ethyl-5-fluoropyrimidine
InChI Formula InChI=1S/C6H6ClFN2/c1-2-4-5(8)6(7)10-3-9-4/h3H,2H2,1H3
SMILES CCc1ncnc(Cl)c1F
IUPAC Name 4-chloro-6-ethyl-5-fluoropyrimidine
Molecular Weight (g/mol) 160.58
Formula Weight 160.0204