Learn More
4-Chloro-5-hydroxy-2-nitrobenzoic Acid Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-5-hydroxy-2-nitrobenzoic Acid Methyl Ester |
| CAS | 1113049-82-3 |
| Molecular Formula | C8H6ClNO5 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C8H6ClNO5/c1-15-8(12)4-2-7(11)5(9)3-6(4)10(13)14/h2-3,11H,1H3 |
| SMILES | COC(=O)c1cc(O)c(Cl)cc1[N+](=O)[O-] |
| IUPAC Name | methyl 4-chloro-5-hydroxy-2-nitrobenzoate |
| Molecular Weight (g/mol) | 231.59 |
| Formula Weight | 230.9935 |