Learn More
(4-Chloro-3-nitrophenyl)-(2-thienyl)methanone, TRC
Specifications
Specifications
| Chemical Name or Material | (4-Chloro-3-nitrophenyl)-(2-thienyl)methanone |
| CAS | 31431-18-2 |
| Molecular Formula | C11 H6 Cl N O3 S |
| Quantity | 10 g |
| InChI Formula | InChI=1S/C11H6ClNO3S/c12-8-4-3-7(6-9(8)13(15)16)11(14)10-2-1-5-17-10/h1-6H |
| SMILES | [O-][N+](=O)c1cc(ccc1Cl)C(=O)c2cccs2 |
| IUPAC Name | (4-chloro-3-nitrophenyl)-thiophen-2-ylmethanone |
| Molecular Weight (g/mol) | 267.69 |
| Formula Weight | 266.9757 |