missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methylphenyl isocyanate, 98%, Thermo Scientific™

Product Code. 11325115
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11325115 10 g 10g
11310436 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11325115 Supplier Thermo Scientific Alfa Aesar Supplier No. H31099.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a intermediates for pharmaceutical.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51488-20-1
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.59
MDL Number MFCD11101408
InChI Key UKTKKMZDESVUEE-UHFFFAOYSA-N
PubChem CID 11400996
SMILES CC1=CC=C(C=C1Cl)N=C=O

Specifications

Quantity 10 g
Refractive Index 1.558
UN Number UN2206
Synonym 4-chloro-3-methylphenyl isocyanate, 4-chloro-3-methylphenylisocyanate, 2-chloro-5-isocyanato toluene, benzene,1-chloro-4-isocyanato-2-methyl, 1-chloranyl-4-isocyanato-2-methyl-benzene, 3-methyl-4-chlorophenyl isocyanate, benzene, 1-chloro-4-isocyanato-2-methyl
Solubility Information Hydrolyzes with water.
IUPAC Name 1-chloro-4-isocyanato-2-methylbenzene
Formula Weight 167.6
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-methylphenyl isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.