Learn More
4-Chloro-3-methyl-2,2-diphenylbutyronitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-3-methyl-2,2-diphenylbutyronitrile |
| CAS | 133132-74-8 |
| Molecular Formula | C17H16ClN |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C17H16ClN/c1-14(12-18)17(13-19,15-8-4-2-5-9-15)16-10-6-3-7-11-16/h2-11,14H,12H2,1H3 |
| SMILES | CC(CCl)C(C#N)(c1ccccc1)c2ccccc2 |
| IUPAC Name | 4-chloro-3-methyl-2,2-diphenylbutanenitrile |
| Molecular Weight (g/mol) | 269.77 |
| Formula Weight | 269.0971 |