Learn More
4-Chloro-3-fluorobenzylamine, 97%, Thermo Scientific™
Product Code. 11386869
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386869 1 g 1g
11396869 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11386869 Supplier Thermo Scientific Alfa Aesar Supplier No. H26780.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 72235-58-6
Molecular Formula C7H7ClFN
Molecular Weight (g/mol) 159.588
MDL Number MFCD04116008
InChI Key HSNPBYKCCNMQNA-UHFFFAOYSA-N
Synonym 4-chloro-3-fluorophenyl methanamine, 4-chloro-3-fluorobenzylamine, 4-chloro-3-fluorobenzyl amine, benzenemethanamine,4-chloro-3-fluoro, benzenemethanamine, 4-chloro-3-fluoro, 4-chloro-3-fluorophenyl methylamine, 1-4-chloro-3-fluorophenyl methanamine, 3-fluoro-4-chlorobenzyl amine, rarechem al bw 1597, 4-chloranyl-3-fluoranyl-phenyl methanamine
PubChem CID 2757541
IUPAC Name (4-chloro-3-fluorophenyl)methanamine
SMILES C1=CC(=C(C=C1CN)F)Cl
Specifications
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 159.59
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-fluorobenzylamine
compliance-icons
Product Identifier
  • 4-Chloro-3-fluorobenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 4-Chloro-3-fluorobenzylamine