missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. p-7028943
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11471270

Brand: Thermo Scientific Alfa Aesar H26360.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 320-43-4
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.564
MDL Number MFCD03092987
InChI Key AHFINSWGYAZBOZ-UHFFFAOYSA-N
Synonym 4-chloro-2-trifluoromethyl benzaldehyde, 2-trifluoromethyl-4-chlorobenzaldehyde, benzaldehyde, 4-chloro-2-trifluoromethyl, pubchem2479, acmc-1ad3e, ksc496c8d, 4-chloro-2-trifluoromethyl-benzaldehyde
PubChem CID 2779017
IUPAC Name 4-chloro-2-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=C(C=C1Cl)C(F)(F)F)C=O

Specifications

Refractive Index 1.496
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 208.57
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-(trifluoromethyl)benzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.