Learn More
4-Chloro-2-isopropyl-6-methylpyrimidine, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-2-isopropyl-6-methylpyrimidine |
| CAS | 4595-69-1 |
| Molecular Formula | C8H11ClN2 |
| Quantity | 10 g |
| Synonym | 4-Chloro-6-methyl-2-(1-methylethyl)pyrimidine |
| InChI Formula | InChI=1S/C8H11ClN2/c1-5(2)8-10-6(3)4-7(9)11-8/h4-5H,1-3H3 |
| SMILES | CC(C)c1nc(C)cc(Cl)n1 |
| IUPAC Name | 4-chloro-6-methyl-2-propan-2-ylpyrimidine |
| Molecular Weight (g/mol) | 170.64 |
| Formula Weight | 170.0611 |