Learn More
4-Chloro-1H-imidazo[4,5-c]pyridine, TRC
Product Code. 30355650
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355650 10 g 10g
30366393 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30355650 Supplier Toronto Research Chemicals Supplier No. TRCC36759510G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2770-01-6
Molecular Formula C11 H16 N5 O . F6 P
Molecular Weight (g/mol) 379.242
Synonym 4-Chloro-3H-imidazo[4,5-c]pyridine, 6-Chloro-3-deazapurine, NSC 611236
IUPAC Name N,N,N',N'-tetramethyl-1-(3-oxidobenzotriazole-1,3-diium-1-ylidene)methanediamine;hexafluorophosphate
SMILES CN(C)C(=[N+]1N=[N+]([O-])c2ccccc12)N(C)C.F[P-](F)(F)(F)(F)F
Specifications
Quantity 10 g
InChI Formula InChI=1S/C11H16N5O.F6P/c1-13(2)11(14(3)4)15-9-7-5-6-8-10(9)16(17)12-15;1-7(2,3,4,5)6/h5-8H,1-4H3;/q+1;-1
Formula Weight 379.1
Chemical Name or Material 4-Chloro-1H-imidazo[4,5-c]pyridine