Learn More
4-Carboxy-9-acridanone (Technical Grade), TRC
Product Code. 30349006
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349006 1 g -
30352606 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30349006 Supplier Toronto Research Chemicals Supplier No. TRCC1777781G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 24782-64-7
Molecular Formula C14H9NO3
Molecular Weight (g/mol) 239.23
Synonym 9,10-Dihydro-9-oxo-4-acridinecarboxylic Acid, 4-Carboxyacridone, 9(10H)-Acridone-4-carboxylic Acid, Acridone-4-carboxylic Acid, 9-oxo-4-acridancarboxylic Acid
IUPAC Name 9-oxo-10H-acridine-4-carboxylic acid
SMILES OC(=O)c1cccc2C(=O)c3ccccc3Nc12
Specifications
Quantity 1 g
InChI Formula InChI=1S/C14H9NO3/c16-13-8-4-1-2-7-11(8)15-12-9(13)5-3-6-10(12)14(17)18/h1-7H,(H,15,16)(H,17,18)
Formula Weight 239.0582
Chemical Name or Material 4-Carboxy-9-acridanone (Technical Grade)