Learn More
4-Bromotoluene-a,a,a-d3, CDN
Specifications
Specifications
| Chemical Name or Material | 4-Bromotoluene-alpha,alpha,alpha-D3 |
| CAS | 22328-44-5 |
| Molecular Formula | C7 D3 H4 Br |
| Quantity | 0.5 g |
| Synonym | Benzene, 1-bromo-4-(methyl-d3)- (9 CI), Toluene-α,α,α-d3, p-bromo- (8 CI), 1-Bromo-4-(2H3)methylbenzene, 1-Bromo-4-(trideuteriomethyl)benzene, p-Bromotoluene-α,α,α-d3, 4-Bromotoluene-α,α,α-d3, 4-Bromotoluene-α,α,α-D3, 4-Bromotoluene-d3, 4-Bromotoluene-D3 (methyl-D3), 1-Bromo-4-(methyl-d3)benzene |
| InChI Formula | InChI=1 S/C7H7Br/c1-6-2-4-7(8)5-3-6/h2-5 H,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])c1ccc(Br)cc1 |
| IUPAC Name | 1-bromo-4-(trideuteriomethyl)benzene |
| Molecular Weight (g/mol) | 174.053 |
| Formula Weight | 172.9919 g/mol |
| Show More |