missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzyl alcohol, 99%

Product Code. p-7021031
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11474614

Brand: Thermo Scientific Alfa Aesar A13197.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 873-75-6 | C7H7BrO | 187.036 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-75-6
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD00004650
InChI Key VEDDBHYQWFOITD-UHFFFAOYSA-N
Synonym 4-bromobenzyl alcohol, 4-bromophenyl methanol, p-bromobenzyl alcohol, benzenemethanol, 4-bromo, para-bromobenzyl alcohol, 4-bromobenzylalcohol, p-bromo-benzyl alcohol, ccris 5119, 4-bromophenyl-methanol, benzyl alcohol, p-bromo
PubChem CID 70119
IUPAC Name (4-bromophenyl)methanol
SMILES C1=CC(=CC=C1CO)Br

Specifications

Melting Point 76°C to 79°C
Quantity 10 g
Beilstein 1931620
Formula Weight 187.04
Percent Purity 99%
Chemical Name or Material 4-Bromobenzyl alcohol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.