Learn More
4-Bromo-3-(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™
Product Code. 11358876
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11358876
Brand: Thermo Scientific Alfa Aesar L19512.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 351003-47-9
Molecular Formula C7H3BrClF3O2S
Molecular Weight (g/mol) 323.508
MDL Number MFCD03094391
InChI Key QCFIMBHKZZLZAE-UHFFFAOYSA-N
Synonym 4-bromo-3-trifluoromethyl benzenesulfonyl chloride, 4-bromo-3-trifluoromethyl benzene-1-sulfonyl chloride, 4-bromo-3-trifluoromethyl benzene sulphonyl chloride, bromo-3-trifluoromethyl benzenesulfonylchloride, 4-bromo-3-trifluoromethyl benzenesulfonylchloride, 4-bromo-3-trifluoromethyl phenyl chlorosulfone, 4-bromo-3-trifluoromethyl benzenesulphonylchloride, 4-bromo-3-trifluoromethyl-benzenesulfonyl chloride, 4-bromo-3-trifluoromethyl benzenesulphonyl chloride, benzenesulfonyl chloride, 4-bromo-3-trifluoromethyl
PubChem CID 2778606
IUPAC Name 4-bromo-3-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)C(F)(F)F)Br
Specifications
Density 1.833
Flash Point >110°C (230°F)
Refractive Index 1.536
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 323.52
Percent Purity 97%
Chemical Name or Material 4-Bromo-3-(trifluoromethyl)benzenesulfonyl chloride