missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-3'-(trifluoromethyl)acetanilide, 97%

Product Code. p-7024743
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11408797 2.5 g 2.5g
11418797 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11408797 Supplier Thermo Scientific Alfa Aesar Supplier No. A19939.88

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 41513-05-7 | C9H7BrF3NO | 282.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41513-05-7
Molecular Formula C9H7BrF3NO
Molecular Weight (g/mol) 282.06
MDL Number MFCD00052361
InChI Key AOHSBSXIGFIMNC-UHFFFAOYSA-N
Synonym n-4-bromo-3-trifluoromethyl phenyl acetamide, 5-acetamido-2-bromobenzotrifluoride, 4-bromo-3-trifluoromethyl acetanilide, 4'-bromo-3'-trifluoromethyl acetanilide, n-4-bromo-3-trifluoromethyl phenyl acet amide, n1-4-bromo-3-trifluoromethyl phenyl acetamide, pubchem2757, maybridge4_003018, buttpark 37\11-74
PubChem CID 697838
IUPAC Name N-[4-bromo-3-(trifluoromethyl)phenyl]acetamide
SMILES CC(=O)NC1=CC(=C(C=C1)Br)C(F)(F)F

Specifications

Melting Point 116°C to 119°C
Quantity 2.5 g
Formula Weight 282.07
Percent Purity 97%
Chemical Name or Material 4'-Bromo-3'-(trifluoromethyl)acetanilide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.