missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-thiophenecarboxylic acid, 97%, Thermo Scientific™

Product Code. p-4509900
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10411334 5 g Amber Glass Bottle 5g
10662685 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10411334 Supplier Thermo Scientific Maybridge Supplier No. CC28501DE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 16694-18-1
Molecular Formula C5H3BrO2S
Molecular Weight (g/mol) 207.041
InChI Key HJZFPRVFLBBAMU-UHFFFAOYSA-N
Synonym 4-bromo-2-thiophenecarboxylic acid, 4-bromo-thiophene-2-carboxylic acid, 2-thiophenecarboxylic acid, 4-bromo, 4-bromothiophene-carboxylic acid, 4-bromo-2-thiophene carboxylic acid, 4-bromo-2-carboxythiophene, 4-bromo-2-thiophenecarboxylicacid, 3-bromothiophene-5-carboxylic acid, pubchem5189, 4-bromo-2-thenoic acid
PubChem CID 610409
IUPAC Name 4-bromothiophene-2-carboxylic acid
SMILES C1=C(SC=C1Br)C(=O)O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 207.05
Percent Purity 95%
Chemical Name or Material 4-Bromo-2-thiophenecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.