Learn More
4-Bromo-2-fluorobenzyl alcohol, 97%, Thermo Scientific Chemicals
Product Code. 11389455
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389455 5 g 5g
11379455 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11389455 Supplier Thermo Scientific Alfa Aesar Supplier No. L18679.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 188582-62-9
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.026
MDL Number MFCD00143265
InChI Key BWBJZMQPVBWEJU-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobenzyl alcohol, 4-bromo-2-fluorophenyl methanol, 4-bromo-2-fluorobenzylalcohol, benzenemethanol, 4-bromo-2-fluoro, rarechem al bd 0386, timtec-bb sbb005826, 4-bromo-2-fluorophenyl methan-1-ol, 2-fluoro-4-bromobenzyl alcohol, pubchem3223, acmc-209erg
PubChem CID 2734027
IUPAC Name (4-bromo-2-fluorophenyl)methanol
SMILES C1=CC(=C(C=C1Br)F)CO
Specifications
Melting Point 31°C to 33°C
Quantity 5 g
Beilstein 7757667
Formula Weight 205.03
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-fluorobenzyl alcohol