missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-2'-fluoroacetanilide, 98%, Thermo Scientific™

Product Code. 11422358
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11422358

Brand: Thermo Scientific Alfa Aesar A15723.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 326-66-9
Molecular Formula C8H7BrFNO
Molecular Weight (g/mol) 232.052
MDL Number MFCD00042588
InChI Key BCYGKMDWQBWUSC-UHFFFAOYSA-N
Synonym n-4-bromo-2-fluorophenyl acetamide, 4'-bromo-2'-fluoroacetanilide, 4-bromo-2-fluoroacetanilide, acetamide, n-4-bromo-2-fluorophenyl, pubchem14886, acmc-1cooc, ksc497m2b, timtec-bb sbb009911, attercop-chm at130693
PubChem CID 67593
IUPAC Name N-(4-bromo-2-fluorophenyl)acetamide
SMILES CC(=O)NC1=C(C=C(C=C1)Br)F

Specifications

Melting Point 154°C to 158°C
Quantity 250 g
Beilstein 3250459
Formula Weight 232.05
Percent Purity 98%
Chemical Name or Material 4'-Bromo-2'-fluoroacetanilide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.