Learn More
4-Benzyloxy-3-methoxybenzyl alcohol, 98%, Thermo Scientific Chemicals
Product Code. 11367289
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367289 5 g 5g
11377289 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11367289 Supplier Thermo Scientific Alfa Aesar Supplier No. H27443.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 33693-48-0
Molecular Formula C15H16O3
Molecular Weight (g/mol) 244.29
MDL Number MFCD00004640
InChI Key PDBXFVPMVYQICB-UHFFFAOYSA-N
Synonym 4-benzyloxy-3-methoxybenzyl alcohol, 4-benzyloxy-3-methoxyphenyl methanol, 4-benzyloxy-3-methoxybenzylalcohol, benzenemethanol, 3-methoxy-4-phenylmethoxy, acmc-20amos, o-benzylvanillyl alcohol, 4-o-benzyl vanillyl alcohol, vanillyl alcohol benzyl ether
PubChem CID 98832
IUPAC Name (3-methoxy-4-phenylmethoxyphenyl)methanol
SMILES COC1=C(C=CC(=C1)CO)OCC2=CC=CC=C2
Specifications
Melting Point 71°C to 73°C
Quantity 5 g
Formula Weight 244.3
Percent Purity 98%
Chemical Name or Material 4-Benzyloxy-3-methoxybenzyl alcohol