Learn More
(4-Azidophenyl)methanol, TRC
Specifications
Specifications
| Chemical Name or Material | (4-Azidophenyl)methanol |
| CAS | 31499-54-4 |
| Molecular Formula | C7 H8 N3 O |
| Quantity | 250 mg |
| Synonym | 4-(Hydroxymethyl)phenyl Azide, 4-Azidobenzyl Alcohol, p-Azidobenzyl Alcohol4-Azidobenzenemethanol, p-Azido-benzyl Alcohol |
| InChI Formula | InChI=1S/C7H8N3O/c8-10-9-7-3-1-6(5-11)2-4-7/h1-4,8,11H,5H2/q+1 |
| SMILES | OCc1ccc(cc1)N=[N+]=N |
| IUPAC Name | [4-(hydroxymethyl)phenyl]imino-iminoazanium |
| Molecular Weight (g/mol) | 150.158 |
| Formula Weight | 150.066 |