Learn More
4'-(Azidomethyl)-[1,1'-biphenyl]-2-carboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4'-(Azidomethyl)-[1,1'-biphenyl]-2-carboxylic Acid |
| CAS | 2758005-15-9 |
| Molecular Formula | C14 H11 N3 O2 |
| Quantity | 50 mg |
| Synonym | 4'-(azidomethyl)-[1,1'-biphenyl]-2-methanoic acid, 2-[4-(azidomethyl)phenyl]benzoic acid, 4'-(Azidomethyl)-[1,1'-biphenyl]-2-carboxylic Acid |
| InChI Formula | InChI=1S/C14H11N3O2/c15-17-16-9-10-5-7-11(8-6-10)12-3-1-2-4-13(12)14(18)19/h1-8H,9H2,(H,18,19) |
| SMILES | OC(=O)c1ccccc1c2ccc(CN=[N+]=[N-])cc2 |
| IUPAC Name | 2-[4-(azidomethyl)phenyl]benzoic acid |
| Molecular Weight (g/mol) | 253.256 |
| Formula Weight | 253.0851 |