Learn More
4-(Azepan-1-yl)benzaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(Azepan-1-yl)benzaldehyde |
| CAS | 50333-45-4 |
| Molecular Formula | C13H17NO |
| Quantity | 500 mg |
| Synonym | p-(Hexamethyleneimino)benzaldehyde, 4-(Hexahydro-1H-azepin-1-yl)benzaldehyde |
| InChI Formula | InChI=1S/C13H17NO/c15-11-12-5-7-13(8-6-12)14-9-3-1-2-4-10-14/h5-8,11H,1-4,9-10H2 |
| SMILES | O=Cc1ccc(cc1)N2CCCCCC2 |
| IUPAC Name | 4-(azepan-1-yl)benzaldehyde |
| Molecular Weight (g/mol) | 203.28 |
| Formula Weight | 203.131 |