Learn More
4-Aminophenyl Alpha-D-Glucopyranoside, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Aminophenyl alpha-D-Glucopyranoside |
| CAS | 31302-52-0 |
| Molecular Formula | C12H17NO6 |
| Quantity | 10 mg |
| Synonym | p-Aminophenyl α-D-Glucopyranoside |
| InChI Formula | InChI=1S/C12H17NO6/c13-6-1-3-7(4-2-6)18-12-11(17)10(16)9(15)8(5-14)19-12/h1-4,8-12,14-17H,5,13H2/t8-,9-,10+,11-,12+/m1/s1 |
| SMILES | OC[C@H]1O[C@H](OC2=CC=C(N)C=C2)[C@H](O)[C@@H](O)[C@@H]1O |
| IUPAC Name | (2R,3R,4S,5S,6R)-2-(4-aminophenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol |
| Molecular Weight (g/mol) | 271.27 |
| Formula Weight | 271.11 |