Learn More
4-Aminophenyl 2-Acetamido-2-deoxy-Beta-D-glucopyranoside, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Aminophenyl 2-Acetamido-2-deoxy-beta-D-glucopyranoside |
| CAS | 14419-59-1 |
| Molecular Formula | C14 H20 N2 O6 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C14H20N2O6/c1-7(18)16-11-13(20)12(19)10(6-17)22-14(11)21-9-4-2-8(15)3-5-9/h2-5,10-14,17,19-20H,6,15H2,1H3,(H,16,18)/t10-,11-,12-,13-,14-/m1/s1 |
| SMILES | CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1Oc2ccc(N)cc2 |
| IUPAC Name | N-[(2S,3R,4R,5S,6R)-2-(4-aminophenoxy)-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide |
| Molecular Weight (g/mol) | 312.32 |
| Formula Weight | 312.1321 |