Learn More
4-Aminophenol-d7, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Aminophenol-d7 |
| CAS | 285132-88-9 |
| Molecular Formula | C6 2H7 N O |
| Quantity | 10 mg |
| Synonym | Phen-2,3,5,6-d4-ol-d, 4-(amino-d2)-, 4-(Amino-d2)phen-2,3,5,6-d4-ol-d, 4-Aminophenol-d7, 4-Aminophenol-D7, p-Aminophenol-d7, 1-(Amino-d2)-4-(hydroxy-d)benzene-2,3,5,6-d4, 4-Amino-1-hydroxybenzene-d7 |
| InChI Formula | InChI=1S/C6H7NO/c7-5-1-3-6(8)4-2-5/h1-4,8H,7H2/i1D,2D,3D,4D/hD3 |
| SMILES | [2H]Oc1c([2H])c([2H])c(N([2H])[2H])c([2H])c1[2H] |
| IUPAC Name | N,N,2,3,5,6-hexadeuterio-4-deuteriooxyaniline |
| Molecular Weight (g/mol) | 116.17 |
| Formula Weight | 116.0967 |