missing translation for 'onlineSavingsMsg'
Learn More

4-Aminocarbonyl-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11480628
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11480628

Brand: Thermo Scientific Alfa Aesar B25178.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87594-59-0
Molecular Formula C8H6N2O5
Molecular Weight (g/mol) 210.145
MDL Number MFCD04039778
InChI Key LKPGLAHLFCDSNK-UHFFFAOYSA-N
Synonym 4-carbamoyl-3-nitrobenzoic acid, 2-nitro-4-carboxybenzamide, 4-carbamoyl-3-nitro-benzoic acid, pubchem13255, 3-nitroterephthalamic acid, 4-carboxy-2-nitrobenzamide, 4-aminocarbonyl-3-nitrobenzoic acid, 4-aminocarbonyl-3-nitro-benzoic acid
PubChem CID 7016425
SMILES C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)N

Specifications

Quantity 1 g
Formula Weight 210.15
Percent Purity 98%
Chemical Name or Material 4-Aminocarbonyl-3-nitrobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.