Learn More
4-Aminobutyric-2,2-d2 Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Aminobutyric acid 2,2-d2 |
| CAS | 67910-98-9 |
| Molecular Formula | C4H7D2NO2 |
| Quantity | 25 mg |
| Synonym | 4-Aminobutyric-2,2-d2 Acid |
| InChI Formula | InChI=1S/C4H9NO2/c5-3-1-2-4(6)7/h1-3,5H2,(H,6,7)/i2D2 |
| SMILES | NCCC([2H])(C(O)=O)[2H] |
| IUPAC Name | 4-aminobutanoic-2,2-d2 acid |
| Molecular Weight (g/mol) | 105.13 |
| Formula Weight | 105.08 |