Learn More
(4-Amino-pyridin-3-yl)-methanol, TRC
Specifications
Specifications
| Chemical Name or Material | (4-Amino-pyridin-3-yl)-methanol |
| CAS | 138116-34-4 |
| Molecular Formula | C6H8N2O |
| Quantity | 250 mg |
| Synonym | 4-Amino-3-pyridinemethanol, 4-Amino-3-hydroxymethylpyridine, 4-Aminopyridine-3-methanol |
| InChI Formula | InChI=1S/C6H8N2O/c7-6-1-2-8-3-5(6)4-9/h1-3,9H,4H2,(H2,7,8) |
| SMILES | OCC1=CNC=CC1=N |
| IUPAC Name | (4-imino-1H-pyridin-3-yl)methanol |
| Molecular Weight (g/mol) | 124.14 |
| Formula Weight | 124.0637 |