Learn More
4-Amino-3-pyridinemethanol, 95%
Product Code. 11444410
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444410 5 g 5g
11434410 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11444410 Supplier Thermo Scientific Alfa Aesar Supplier No. H50033.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 138116-34-4 | C6H8N2O | 124.143 g/mol
4-Amino-3-pyridinemethanol is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Amino-3-pyridinemethanol is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 138116-34-4
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD06203066
InChI Key WNBVEYMTVDMSFZ-UHFFFAOYSA-N
Synonym 4-aminopyridin-3-yl methanol, 4-amino-pyridin-3-yl-methanol, 4-aminopyridine-3-methanol, 4-amino-3-pyridinemethanol, 3-pyridinemethanol, 4-amino, 4-amino-3-hydroxymethylpyridine, 4-amino-3-pyridyl methan-1-ol, 4-amino-3-pyridinyl methanol, rarechem al bd 1386, acmc-1c1ii
PubChem CID 10796739
IUPAC Name (4-aminopyridin-3-yl)methanol
SMILES C1=CN=CC(=C1N)CO
Specifications
Melting Point 167°C to 169°C
Quantity 5 g
Sensitivity Air sensitive
Solubility Information Slightly soluble in water.
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material 4-Amino-3-pyridinemethanol