Learn More
(4-Amino-3-nitrophenyl)-(2-thienyl)methanon, TRC
Specifications
Specifications
| Chemical Name or Material | (4-Amino-3-nitrophenyl)-(2-thienyl)methanone |
| CAS | 31431-30-8 |
| Molecular Formula | C11 H8 N2 O3 S |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C11H8N2O3S/c12-8-4-3-7(6-9(8)13(15)16)11(14)10-2-1-5-17-10/h1-6H,12H2 |
| SMILES | Nc1ccc(cc1[N+](=O)[O-])C(=O)c2cccs2 |
| IUPAC Name | (4-amino-3-nitrophenyl)-thiophen-2-ylmethanone |
| Molecular Weight (g/mol) | 248.26 |
| Formula Weight | 248.0256 |