missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,6-dihydroxypyrimidine, 98%

Product Code. p-55702 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Pakningsstørrelse:
100g
25g
500g
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 10433585

Brand: Thermo Scientific Acros 103520250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

CAS: 873-83-6 | C4H5N3O2 | 127.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 873-83-6
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00006071
InChI Key LNDZXOWGUAIUBG-UHFFFAOYSA-N
Synonym 6-aminouracil, 4-amino-2,6-dihydroxypyrimidine, 6-aminopyrimidine-2,4 1h,3h-dione, 4-aminouracil, 6-aminopyrimidine-2,4-diol, 6-amino-2,4-pyrimidinediol, 2,4 1h,3h-pyrimidinedione, 6-amino, uracil, 6-amino, cytosine, 6-hydroxy, 6-amino-2,4-dihydroxypyrimidine
PubChem CID 70120
SMILES NC1=CC(=O)NC(=O)N1

Tekniske data

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color Brown to Cream
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Quantity 25 g
Beilstein 24, 469
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in methanol
IUPAC Name 6-amino-1H-pyrimidine-2,4-dione
Formula Weight 127.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2, 6-dihydroxypyrimidine, 98%
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • 4-Amino-2,6-dihydroxypyrimidine
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sikkerhedssætning
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.