Learn More
4-Amino-2,2,6,6-tetramethylpiperidinyloxy, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Amino-2,2,6,6-tetramethylpiperidinyloxy |
| CAS | 14691-88-4 |
| Molecular Formula | C9 H19 N2 O |
| Quantity | 10 g |
| Synonym | 1-Piperidinyloxy, 4-amino-2,2,6,6-tetramethyl- (9CI, ACI), 4-Amino-2,2,6,6-tetramethyl-1-piperidinyloxy (ACI), Piperidinooxy, 4-amino-2,2,6,6-tetramethyl- (8CI), (2,2,6,6-Tetramethyl-1-oxy-4-piperidinyl)amine, 2,2,6,6-Tetramethyl-1-oxy-4-aminopiperidine, 2,2,6,6-Tetramethyl-4-amino-1-piperidinyloxy, 2,2,6,6-Tetramethyl-4-aminopiperidine N-oxide, 2,2,6,6-Tetramethyl-4-aminopiperidine-1-oxyl, 4-Amino-1-oxyl-2,2,6,6-tetramethylpiperidine, 4-Amino-2,2,6,6-tetramethyl-1-piperidinyloxyl, 4-Amino-2,2,6,6-tetramethylpiperidin-1-oxyl, 4-Amino-2,2,6,6-tetramethylpiperidin-N-oxyl, 4-Amino-2,2,6,6-tetramethylpiperidine N-oxide, 4-Amino-2,2,6,6-tetramethylpiperidine-1-nitroxide, 4-Amino-2,2,6,6-tetramethylpiperidine-1-oxy, 4-Amino-2,2,6,6-tetramethylpiperidine-1-oxyl, 4-Amino-2,2,6,6-tetramethylpiperidine-N-oxyl, 4-Amino-2,2,6,6-tetramethylpiperidino-1-oxy, 4-Amino-2,2,6,6-tetramethylpiperidino-1-oxyl, 4-Amino-2,2,6,6-tetramethylpiperidinooxyl, 4-Amino-2,2,6,6-tetramethylpiperidinyl-N-oxy, 4-Amino-2,2,6,6-tetramethylpiperidinyloxy, 4-Aminotempo, 4-NH2-TEMPO, 6-Tempamine, ST 7820, TAMINE, Tempamine, Tempo-amine |
| InChI Formula | InChI=1S/C9H19N2O/c1-8(2)5-7(10)6-9(3,4)11(8)12/h7H,5-6,10H2,1-4H3 |
| SMILES | CC1(C)CC(N)CC(C)(C)N1[O] |
| IUPAC Name | 1-λ^{1}-oxidanyl-2,2,6,6-tetramethylpiperidin-4-amine |
| Molecular Weight (g/mol) | 171.26 |
| Formula Weight | 171.1497 |