Learn More
4-Amino-1-naphthalenesulfonic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Amino-1-naphthalenesulfonic Acid |
| CAS | 84-86-6 |
| Molecular Formula | C10 H9 N O3 S |
| Quantity | 10 g |
| Synonym | Naphthionic Acid, 1-Naphthalenesulfonic acid, 4-amino-, 4-Amino-1-naphthalenesulfonic acid, 1,4-Naphthionic acid, 1-Amino-4-naphthalenesulfonic acid, 1-Amino-4-sulfonaphthalene, 1-Naphthylamine-4-sulfonic acid, 4-Aminonaphthalenesulfonic acid, NSC 4155, Naphthionic acid, Piria's acid, α-Naphthylamine-4-sulfonic acid, Naphthionic Acid, 1-Naphthalenesulfonic acid, 4-amino-, 4-Amino-1-naphthalenesulfonic acid, 1,4-Naphthionic acid, 1-Amino-4-naphthalenesulfonic acid, 1-Amino-4-sulfonaphthalene, 1-Naphthylamine-4-sulfonic acid, 4-Aminonaphthalenesulfonic acid, NSC 4155, Naphthionic acid, Piria's acid, α-Naphthylamine-4-sulfonic acid |
| InChI Formula | InChI=1S/C10H9NO3S/c11-9-5-6-10(15(12,13)14)8-4-2-1-3-7(8)9/h1-6H,11H2,(H,12,13,14) |
| SMILES | Nc1ccc(c2ccccc12)S(=O)(=O)O |
| IUPAC Name | 4-aminonaphthalene-1-sulfonic acid |
| Molecular Weight (g/mol) | 223.25 |
| Formula Weight | 223.0303 |