Learn More
4-(Acetyloxy)-3-methoxybenzenethanol Acetate, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(Acetyloxy)-3-methoxybenzenethanol Acetate |
| CAS | 32022-28-9 |
| Molecular Formula | C13H16O5 |
| Quantity | 100 mg |
| Synonym | 4-(Acetyloxy)-3-methoxybenzeneethanol 1-Acetate, 4-Hydroxy-3-methoxyphenethyl Alcohol Diacetate |
| InChI Formula | InChI=1S/C13H16O5/c1-9(14)17-7-6-11-4-5-12(18-10(2)15)13(8-11)16-3/h4-5,8H,6-7H2,1-3H3 |
| SMILES | COc1cc(CCOC(=O)C)ccc1OC(=O)C |
| IUPAC Name | 2-(4-acetyloxy-3-methoxyphenyl)ethyl acetate |
| Molecular Weight (g/mol) | 252.26 |
| Formula Weight | 252.0998 |