Learn More
4-(Acetyloxy)-3-methoxybenzenethanol Acetate, TRC
Product Code. 30326100
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326100 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30326100 Supplier Toronto Research Chemicals Supplier No. TRCA1675801G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-(Acetyloxy)-3-methoxybenzenethanol Acetate
CAS 32022-28-9
Molecular Formula C13H16O5
Quantity 1 g
Synonym 4-(Acetyloxy)-3-methoxybenzeneethanol 1-Acetate, 4-Hydroxy-3-methoxyphenethyl Alcohol Diacetate
InChI Formula InChI=1S/C13H16O5/c1-9(14)17-7-6-11-4-5-12(18-10(2)15)13(8-11)16-3/h4-5,8H,6-7H2,1-3H3
SMILES COc1cc(CCOC(=O)C)ccc1OC(=O)C
IUPAC Name 2-(4-acetyloxy-3-methoxyphenyl)ethyl acetate
Molecular Weight (g/mol) 252.26
Formula Weight 252.0998