Learn More
4,7-Dihydro-4-oxo-2-[(triphenylmethyl)amino]-3H-pyrrolo[2,3-d]pyrimidine-5-carbonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 4,7-Dihydro-4-oxo-2-[(triphenylmethyl)amino]-3H-pyrrolo[2,3-d]pyrimidine-5-carbonitrile |
| CAS | 1019853-64-5 |
| Molecular Formula | C26H19N5O |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C26H19N5O/c27-16-18-17-28-23-22(18)24(32)30-25(29-23)31-26(19-10-4-1-5-11-19,20-12-6-2-7-13-20)21-14-8-3-9-15-21/h1-15,17H,(H3,28,29,30,31,32) |
| SMILES | O=C1C2=C(NC=C2C#N)N=C(NC(C3=CC=CC=C3)(C4=CC=CC=C4)C5=CC=CC=C5)N1 |
| IUPAC Name | 4-oxo-2-(tritylamino)-4,7-dihydro-3H-pyrrolo[2,3-d]pyrimidine-5-carbonitrile |
| Molecular Weight (g/mol) | 417.46 |
| Formula Weight | 417.16 |