Learn More
4,6-Dimethoxy-2'-methyl-3,4',6'-grisantrione, TRC
Specifications
Specifications
| Chemical Name or Material | 4,6-Dimethoxy-2'-methyl-3,4',6'-grisantrione |
| CAS | 2855-92-7 |
| Molecular Formula | C16H16O6 |
| Quantity | 10 mg |
| Synonym | 4,6-Dimethoxy-6'-methyl-spiro[benzofuran-2(3H),1'-cyclohexane]-2',3,4'-trione |
| InChI Formula | InChI=1S/C16H16O6/c1-8-4-9(17)5-13(18)16(8)15(19)14-11(21-3)6-10(20-2)7-12(14)22-16/h6-8H,4-5H2,1-3H3 |
| SMILES | COc1cc(OC)c2C(=O)C3(Oc2c1)C(C)CC(=O)CC3=O |
| IUPAC Name | 4,6-dimethoxy-5'-methylspiro[1-benzofuran-2,4'-cyclohexane]-1',3,3'-trione |
| Molecular Weight (g/mol) | 304.29 |
| Formula Weight | 304.0947 |