missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Trifluoromethyl-2-pyridyloxy)benzamidoxime, Thermo Scientific™

Product Code. 11440200
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440200 1 g 1g
11430200 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440200 Supplier Thermo Scientific Alfa Aesar Supplier No. H50662.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 263161-34-8
Molecular Formula C13H10F3N3O2
Molecular Weight (g/mol) 297.237
MDL Number MFCD08060954
InChI Key RHAVFJMKHOLIST-UHFFFAOYSA-N
Synonym n-hydroxy-4-5-trifluoromethyl pyridin-2-yl oxybenzenecarboximidamide, n-hydroxy-4-5-trifluoromethyl pyridin-2-yl oxy benzimidamide, z-n'-hydroxy-4-5-trifluoromethyl pyridin-2-yl oxy benzene-1-carboximidamide, e-n'-hydroxy-4-5-trifluoromethyl pyridin-2-yl oxy benzenecarboximidamide, n'-hydroxy-4-5-trifluoromethyl pyridin-2-yl oxy benzenecarboximidamide, 4-5-trifluoromethyl-2-pyridyloxy benzamidoxime, hydroxytrifluoromethylpyridinyloxybenzenecarboximidamide, z-n'-hydroxy-4-5-trifluoromethyl pyridin-2-yl oxy benzimidamide
PubChem CID 9582381
IUPAC Name N'-hydroxy-4-[5-(trifluoromethyl)pyridin-2-yl]oxybenzenecarboximidamide
SMILES C1=CC(=CC=C1C(=NO)N)OC2=NC=C(C=C2)C(F)(F)F

Specifications

Melting Point 160°C to 163°C
Quantity 1 g
Formula Weight 297.24
Chemical Name or Material 4-(5-Trifluoromethyl-2-pyridyloxy)benzamidoxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.