Learn More
4-(5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl)benzoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl)benzoic Acid |
| CAS | 189365-92-2 |
| Molecular Formula | C9H6N2O4 |
| Quantity | 50 mg |
| Synonym | Benzoic acid','4-(2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl)- (9CI','ACI), 4-(2,5-Dihydro-5-oxo-1,2,4-oxadiazol-3-yl)benzoic acid (ACI), 4-(5-Oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl)benzoic acid |
| InChI Formula | InChI=1S/C9H6N2O4/c12-8(13)6-3-1-5(2-4-6)7-10-9(14)15-11-7/h1-4H,(H,12,13)(H,10,11,14) |
| SMILES | OC(=O)c1ccc(cc1)C2=NC(=O)ON2 |
| IUPAC Name | 4-(5-oxo-2H-1,2,4-oxadiazol-3-yl)benzoic acid |
| Molecular Weight (g/mol) | 206.15 |
| Formula Weight | 206.0328 |