Learn More
4,5-Epoxy-13-ethyl-17-hydroxy-18,19-dinor-17Alpha-pregn-20-yn-3-one, TRC
Product Code. 30369693
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369693 25 mg
30368800 100 mg
30378787 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30369693 Supplier Toronto Research Chemicals Supplier No. TRCE89194025MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25073-30-7
Molecular Formula C21H28O3
Molecular Weight (g/mol) 328.45
Synonym Levonorgestrel Impurity 7 (Mixture of Diastereomers)
SMILES CC[C@]12CC[C@H]3[C@@H](CCC45OC4C(=O)CC[C@H]35)[C@@H]1CC[C@@]2(O)C#C
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C21H28O3/c1-3-19-10-7-14-13(15(19)9-11-20(19,23)4-2)8-12-21-16(14)5-6-17(22)18(21)24-21/h2,13-16,18,23H,3,5-12H2,1H3/t13-,14+,15+,16-,18?,19+,20+,21?/m1/s1
Formula Weight 328.2038
Chemical Name or Material 4,5-Epoxy-13-ethyl-17-hydroxy-18,19-dinor-17alpha-pregn-20-yn-3-one