Learn More
4,5-Dihydro-2-methoxy-1H-imidazole, (30-40% solution in Dichloromethane), TRC
Product Code. 30372499
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372499 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30372499 Supplier Toronto Research Chemicals Supplier No. TRCD4491550.1ML
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4,5-Dihydro-2-methoxy-1H-imidazole
CAS 28118-54-9
Molecular Formula C4 H8 N2 O
Quantity 0.1 mL
Synonym 1H-Imidazole, 4,5-dihydro-2-methoxy- (9CI, ACI), 2-Imidazoline, 2-methoxy- (6CI, 8CI), 4,5-Dihydro-2-methoxy-1H-imidazole (ACI), 2-Methoxy-2-imidazoline, 2-Methoxyimidazoline, Lys Tag 4H, 2-Methoxy-4,5-dihydro-1H-imidazole
InChI Formula InChI=1S/C4H8N2O/c1-7-4-5-2-3-6-4/h2-3H2,1H3,(H,5,6)
SMILES COC1=NCCN1
IUPAC Name 2-methoxy-4,5-dihydro-1H-imidazole
Molecular Weight (g/mol) 100.119
Formula Weight 100.064
Concentration or Composition (by Analyte or Components) 30-40 %