Learn More
4,5-Dihydro-2-methoxy-1H-imidazole, (30-40% solution in Dichloromethane), TRC
Specifications
Specifications
| Chemical Name or Material | 4,5-Dihydro-2-methoxy-1H-imidazole |
| CAS | 28118-54-9 |
| Molecular Formula | C4 H8 N2 O |
| Quantity | 1 mL |
| Synonym | 1H-Imidazole, 4,5-dihydro-2-methoxy- (9CI, ACI), 2-Imidazoline, 2-methoxy- (6CI, 8CI), 4,5-Dihydro-2-methoxy-1H-imidazole (ACI), 2-Methoxy-2-imidazoline, 2-Methoxyimidazoline, Lys Tag 4H, 2-Methoxy-4,5-dihydro-1H-imidazole |
| InChI Formula | InChI=1S/C4H8N2O/c1-7-4-5-2-3-6-4/h2-3H2,1H3,(H,5,6) |
| SMILES | COC1=NCCN1 |
| IUPAC Name | 2-methoxy-4,5-dihydro-1H-imidazole |
| Molecular Weight (g/mol) | 100.119 |
| Formula Weight | 100.064 |
| Show More |