missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-2-methylimidazole, 97%

Product Code. p-7025272
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499117 1 g 1g
11409127 5 g 5g
11419127 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499117 Supplier Thermo Scientific Alfa Aesar Supplier No. B20667.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15965-33-0 | C4H4Cl2N2 | 150.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15965-33-0
Molecular Formula C4H4Cl2N2
Molecular Weight (g/mol) 150.99
MDL Number MFCD00051639
InChI Key ZWRYWVBALYOSPQ-UHFFFAOYSA-N
Synonym 4,5-dichloro-2-methylimidazole, 1h-imidazole,2-methyl,4,5-dichloro, acmc-1bt3e, 4.5-dichloro-2-methylimidazole, 1h-imidazole,4,5-dichloro-2-methyl, #
PubChem CID 312390
IUPAC Name 4,5-dichloro-2-methyl-1H-imidazole
SMILES CC1=NC(Cl)=C(Cl)N1

Specifications

Melting Point ∼248°C (decomposition)
Quantity 1 g
Beilstein 606442
Formula Weight 151
Percent Purity 97%
Chemical Name or Material 4,5-Dichloro-2-methylimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.