missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Morpholinyl)benzonitrile, 98%, Thermo Scientific™

Catalog Number p-7054076
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
10282-31-2
C11H12N2O
188.23
MFCD00100115
ZSCUWVQXQDCSRV-UHFFFAOYSA-N
394808
4-morpholin-4-ylbenzonitrile
C1COCCN1C2=CC=C(C=C2)C#N
This item is not returnable. View return policy
10282-31-2
C11H12N2O
188.23
MFCD00100115
ZSCUWVQXQDCSRV-UHFFFAOYSA-N
394808
4-morpholin-4-ylbenzonitrile
C1COCCN1C2=CC=C(C=C2)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 85°C
Color White to Brown
Quantity 1 g
UN Number UN3439
Synonym 4-morpholinobenzonitrile, 4-4-cyanophenyl morpholine, 4-morpholin-4-yl benzonitrile, 4-4-morpholinyl benzonitrile, 4-morpholin-4-yl-benzonitrile, benzonitrile, 4-4-morpholinyl, benzonitrile,4-4-morpholinyl, 4-morpholin-4-ylbenzenecarbonitrile, pubchem18708, acmc-1bqst
Solubility Information Soluble in water.
Formula Weight 188.23
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4-(4-Morpholinyl)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'