Learn More
4-[(4-Fluorophenyl)carbonyl]benzonitrile, TRC
Product Code. 30404982
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30404982 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30404982 Supplier Toronto Research Chemicals Supplier No. TRCF621315250MG
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-[(4-Fluorophenyl)carbonyl]benzonitrile
CAS 54978-50-6
Molecular Formula C14H8FNO
Quantity 250 mg
Synonym 4-(4-Fluorobenzoyl)benzonitrile
InChI Formula InChI=1S/C14H8FNO/c15-13-7-5-12(6-8-13)14(17)11-3-1-10(9-16)2-4-11/h1-8H
SMILES O=C(C1=CC=C(C#N)C=C1)C2=CC=C(F)C=C2
IUPAC Name 4-(4-fluorobenzoyl)benzonitrile
Molecular Weight (g/mol) 225.22
Formula Weight 225.06