Learn More
4-(4-Fluorophenyl)aniline, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(4-Fluorophenyl)aniline |
| CAS | 324-93-6 |
| Molecular Formula | C12H10FN |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C12H10FN/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8H,14H2 |
| SMILES | Nc1ccc(cc1)c2ccc(F)cc2 |
| Molecular Weight (g/mol) | 187.21 |
| Formula Weight | 187.0797 |