Learn More
4,4-Dimethyloctane, 99%, Thermo Scientific Chemicals
Product Code. 11407144
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11407144 5 g 5g
11497134 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.
We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 11407144 Supplier Thermo Scientific Alfa Aesar Supplier No. B21554.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.
We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15869-95-1
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00048813
InChI Key ZMEDGZAGMLTROM-UHFFFAOYSA-N
Synonym 4,4-dimethyl octane, octane, 4,4-dimethyl, acmc-1bqkk
PubChem CID 85151
IUPAC Name 4,4-dimethyloctane
SMILES CCCCC(C)(C)CCC
Specifications
Density 0.74
Boiling Point 161°C
Quantity 5 g
UN Number UN3295
Formula Weight 142.29
Percent Purity 99%
Chemical Name or Material 4,4-Dimethyloctane