Learn More
4,4-Dimethyl-2-(4-bromophenyl)-2-oxazoline, TRC
Specifications
Specifications
| Chemical Name or Material | 4,4-Dimethyl-2-(4-bromophenyl)-2-oxazoline |
| CAS | 32664-14-5 |
| Molecular Formula | C11H12BrNO |
| Quantity | 25 mg |
| Synonym | 2-(4-Bromophenyl)-4,4-dimethyl-2-oxazoline, 2-(4-Bromophenyl)-4,4-dimethyl-4,5-dihydro-1,3-oxazole, 2-(4-Bromophenyl)-4,4-dimethyl-4,5-dihydrooxazole, 2-(4-Bromophenyl)-4,4-dimethyloxazoline, 2-(4-Bromophenyl)-4,5-dihydro-4,4-dimethyloxazole, 2-(p-Bromophenyl)-4,4-dimethyl-2-oxazoline |
| InChI Formula | InChI=1S/C11H12BrNO/c1-11(2)7-14-10(13-11)8-3-5-9(12)6-4-8/h3-6H,7H2,1-2H3 |
| SMILES | CC1(C)COC(=N1)c2ccc(Br)cc2 |
| IUPAC Name | 2-(4-bromophenyl)-4,4-dimethyl-5H-1,3-oxazole |
| Molecular Weight (g/mol) | 254.12 |
| Formula Weight | 253.0102 |