missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride, 97%

Product Code. p-90652 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10580011 1 g Glass bottle 1g
10625722 5 g Glass bottle 5g
10625912 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10580011 Supplier Thermo Scientific Acros Supplier No. 109930010
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51304-61-1 | C11H12ClN·HCl | 230.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51304-61-1
MDL Number MFCD00012753
InChI Key NGDJUNABWNTULX-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl-1,2,3,6-tetrahydropyridine hydrochloride, 4-4-chlorophenyl-1,2,3,6-tetrahydropyridinium chloride, 4-4-chlorophenyl-1,2,5,6-tetrahydropyridine, chloride, acmc-1alu0, 4-4-chlorophenyl-1,2,3,6-tetrahydropyridine hcl, 4-4-chloro-phenyl-1,2,3,6-tetrahydro-pyridine hydrochloride, 4-4-chlorophenyl-1?2?3?6-tetrahydropyridine hydrochloride, 4-4-chlorophenyl-1,2,3,6-tetrahydropyridine monohydrochloride, pyridine, 4-4-chlorophenyl-1,2,3,6-tetrahydro-, hydrochloride
PubChem CID 2723612
IUPAC Name 4-(4-chlorophenyl)-1,2,3,6-tetrahydropyridine;hydrochloride
SMILES C1CNCC=C1C2=CC=C(C=C2)Cl.Cl

Specifications

Melting Point 199.0°C to 204.0°C
CAS Min % 96.0
Color White to Yellow
CAS Max % 100.0
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 97%
Molecular Formula C11H12ClN·HCl
Molecular Weight (g/mol) 230.14
Formula Weight 230.14
Percent Purity 97%
Chemical Name or Material 4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.